Assay Details
| Gene Symbol: Rab7l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133590.1 | NP_598274.1 | 2 - 3 | 537 | 80 | |
| XM_006249744.2 | XP_006249806.1 | 3 - 4 | 514 | 80 | ||
| XM_006249745.2 | XP_006249807.1 | 3 - 4 | 371 | 80 | ||
| GenBank mRNA | BC081732.1 | - | 3 - 4 | 493 | 7109012 | 80 |
| FQ234855.1 | - | 2 - 3 | 303 | 80 | ||
| X96663.1 | - | 2 - 3 | 537 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab7l1
Genomic Map