Assay Details
| Gene Symbol: Slc12a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134363.1 | NP_599190.1 | 6 - 7 | 731 | 79 | |
| GenBank mRNA | EF641113.1 | - | 744 | 79 | ||
| U55816.1 | - | 6 - 7 | 731 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc12a5
Genomic Map