Assay Details
| Gene Symbol: Ipmk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134417.1 | NP_599244.1 | 1 - 2 | 256 | 69 | |
| XM_006256345.2 | XP_006256407.1 | 1 - 2 | 309 | 69 | ||
| GenBank mRNA | AY014898.1 | - | 1 - 2 | 256 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P367798 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62553 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P367798 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P367798 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P367798 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P367798 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82753 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 95178 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 993 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10832684 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832687 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832682 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832683 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832686 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832681 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832685 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10832680 | - | - |
| Rat Genome 230 2.0 Array | 1368340_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375801_at | - | - |
| Rat Genome 230 2.0 Array | 1375801_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368340_at | - | - |
| Affymetrix HT Rat Focus Array | 1375801_at | - | - |
| Affymetrix HT Rat Focus Array | 1375801_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368340_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368340_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR2000011514.rn.1 | TC2000001392.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000011512.rn.1 | TC2000001392.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000011516.rn.1 | TC2000001392.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368340_at | - | - |
| Rat Expression Set 230 | 1368340_PM_at | - | - |
| Rat Expression Set 230 | 1375801_PM_at | - | - |
| Rat Expression Set 230 | 1375801_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832680 | - | - |
| Rat Genome U34 Set | rc_AI013446_at | - | - |
| Rat Genome U34 Set | rc_AA997714_at | - | - |
| Rat Genome U34 Set | rc_AI171920_s_at | - | - |
| Rat Genome U34 Set | rc_AI070823_at | - | - |
| Rat Genome U34 Set | rc_AI169399_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21331086 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355219 | - | - |
| RatRef-12 Expression BeadChip | ILMN_65122 | - | - |

Ipmk
Genomic Map