Assay Details
| Gene Symbol: Stau2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007149.1 | NP_001007150.1 | 5 - 6 | 1069 | 135 | |
| NM_001007150.1 | NP_001007151.1 | 3 - 4 | 796 | 135 | ||
| NM_134466.2 | NP_604461.2 | 4 - 5 | 892 | 135 | ||
| GenBank mRNA | AF483620.1 | - | 5 - 6 | 1069 | 135 | |
| AF483621.1 | - | 5 - 6 | 1069 | 135 | ||
| AY549445.1 | - | 3 - 4 | 796 | 135 | ||
| AY549446.1 | - | 3 - 4 | 796 | 135 | ||
| AY549447.1 | - | 4 - 5 | 892 | 135 | ||
| AY549448.1 | - | 4 - 5 | 892 | 135 | ||
| AY684789.1 | - | 796 | 135 | |||
| AY684790.1 | - | 796 | 135 | |||
| BC085705.1 | - | 1428 | 7190916 | 135 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P117984 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P534028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34997 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P534028 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P117984 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P117984 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P534028 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P534028 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P117984 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P542909 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P447440 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P715863 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P534028 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45097 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93253 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49542 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 25233 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 61994 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20488 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81411 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80722 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10940613 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10940609 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10940612 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10940609 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10940613 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10940612 | - | - |
| Rat Genome 230 2.0 Array | 1370680_at | - | - |
| Rat Genome 230 2.0 Array | 1390558_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379754_at | - | - |
| Rat Genome 230 2.0 Array | 1379754_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387456_at | - | - |
| Rat Genome 230 2.0 Array | 1390558_at | - | - |
| Rat Genome 230 2.0 Array | 1370680_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387456_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370680_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387456_at | - | - |
| Affymetrix HT Rat Focus Array | 1390558_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1390558_at | - | - |
| Affymetrix HT Rat Focus Array | 1379754_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387456_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379754_at | - | - |
| Affymetrix HT Rat Focus Array | 1370680_at | - | - |
| Rat Expression Set 230 | 1370680_at | - | - |
| Rat Expression Set 230 | 1390558_PM_at | - | - |
| Rat Expression Set 230 | 1387456_at | - | - |
| Rat Expression Set 230 | 1370680_PM_at | - | - |
| Rat Expression Set 230 | 1387456_PM_at | - | - |
| Rat Expression Set 230 | 1390558_at | - | - |
| Rat Expression Set 230 | 1379754_PM_at | - | - |
| Rat Expression Set 230 | 1379754_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10940613 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10940612 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10940609 | - | - |
| Rat Genome U34 Set | rc_AI070598_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21927890 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376567 | - | - |

Stau2
Genomic Map