Assay Details
| Gene Symbol: Prl2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138861.1 | NP_620216.1 | 1 - 2 | 80 | 71 | |
| GenBank mRNA | AB022882.1 | - | 1 - 2 | 77 | 71 | |
| AF234635.1 | - | 1 - 2 | 80 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB022882.1, AF234635.1, NM_138861.1 transcripts.
Set Membership: |
|

Prl2b1
Genomic Map