Assay Details
| Gene Symbol: Cep19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138865.1 | NP_620220.1 | 1 - 2 | 141 | 75 | |
| GenBank mRNA | AF146738.1 | - | 1 - 2 | 141 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF146738.1, NM_138865.1 transcripts.
Set Membership: |
|

Cep19
Genomic Map