Assay Details
| Gene Symbol: Csnk1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139060.3 | NP_620691.2 | 7 - 8 | 1370 | 56 | |
| XM_008768511.1 | XP_008766733.1 | 7 - 8 | 1375 | 56 | ||
| GenBank mRNA | AB063114.1 | - | 7 - 8 | 1362 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cell cycle
positive regulation of protein phosphorylation
protein phosphorylation
endocytosis
regulation of cell shape
Wnt signaling pathway
peptidyl-serine phosphorylation
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
circadian regulation of gene expression
regulation of circadian rhythm
spindle assembly
positive regulation of canonical Wnt signaling pathway
glycoprotein binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
peptide binding
tau-protein kinase activity
phosphoprotein binding
mitotic cell cycle
positive regulation of protein phosphorylation
protein phosphorylation
endocytosis
regulation of cell shape
Wnt signaling pathway
peptidyl-serine phosphorylation
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
circadian regulation of gene expression
regulation of circadian rhythm
spindle assembly
positive regulation of canonical Wnt signaling pathway
glycoprotein binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
peptide binding
tau-protein kinase activity
phosphoprotein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P133899 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P133904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176579 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P133899 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P133904 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P133904 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P133899 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P133899 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P133904 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P994293 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P509473 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 8255 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93240 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 65387 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10749754 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749752 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749745 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749746 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749749 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749755 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749751 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749753 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749750 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749756 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749748 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10749742 | - | - |
| Rat Genome 230 2.0 Array | 1370819_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1395914_at | - | - |
| Rat Genome 230 2.0 Array | 1370528_at | - | - |
| Rat Genome 230 2.0 Array | 1370527_a_at | - | - |
| Rat Genome 230 2.0 Array | 1373332_at | - | - |
| Rat Genome 230 2.0 Array | 1370819_at | - | - |
| Rat Genome 230 2.0 Array | 1373332_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370527_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1370528_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1395914_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395914_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370527_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1370528_at | - | - |
| Affymetrix HT Rat Focus Array | 1370528_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370819_at | - | - |
| Affymetrix HT Rat Focus Array | 1370527_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1370819_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395914_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000039324.rn.1 | TC1000004300.rn.1 | RTA TC |
| Rat Expression Set 230 | 1373332_at | - | - |
| Rat Expression Set 230 | 1370527_a_at | - | - |
| Rat Expression Set 230 | 1370819_PM_at | - | - |
| Rat Expression Set 230 | 1370527_PM_a_at | - | - |
| Rat Expression Set 230 | 1373332_PM_at | - | - |
| Rat Expression Set 230 | 1370819_at | - | - |
| Rat Expression Set 230 | 1370528_at | - | - |
| Rat Expression Set 230 | 1370528_PM_at | - | - |
| Rat Expression Set 230 | 1395914_at | - | - |
| Rat Expression Set 230 | 1395914_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10749742 | - | - |
| Rat Genome U34 Set | L07578_at | - | - |
| Rat Genome U34 Set | rc_AI013513_at | - | - |
| Rat Genome U34 Set | rc_AA946432_at | - | - |
| Rat Genome U34 Set | rc_AI013513_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20732713 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371863 | - | - |
| RatRef-12 Expression BeadChip | ILMN_57451 | - | - |

Csnk1d
Genomic Map