Assay Details
| Gene Symbol: Cxcl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139089.1 | NP_620789.1 | 1 - 2 | 158 | 58 | |
| GenBank mRNA | BC058444.1 | - | 1 - 2 | 173 | 6918440 | 58 |
| FQ225217.1 | - | 1 - 2 | 172 | 58 | ||
| FQ225221.1 | - | 1 - 2 | 172 | 58 | ||
| FQ225582.1 | - | 1 - 2 | 169 | 58 | ||
| FQ225670.1 | - | 1 - 2 | 172 | 58 | ||
| FQ226479.1 | - | 1 - 2 | 169 | 58 | ||
| FQ232883.1 | - | 1 - 2 | 172 | 58 | ||
| FQ233623.1 | - | 1 - 2 | 172 | 58 | ||
| FQ233884.1 | - | 1 - 2 | 172 | 58 | ||
| U17035.1 | - | 1 - 2 | 158 | 58 | ||
| U22520.1 | - | 1 - 2 | 157 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cxcl10
Genomic Map