Assay Details
| Gene Symbol: Atp5e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139099.1 | NP_620799.1 | 2 - 3 | 227 | 72 | |
| GenBank mRNA | AF010323.1 | - | 2 - 3 | 227 | 72 | |
| BC058133.1 | - | 2 - 3 | 217 | 6887013 | 72 | |
| FQ211630.1 | - | 2 - 3 | 249 | 72 | ||
| FQ211898.1 | - | 2 - 3 | 261 | 72 | ||
| FQ214112.1 | - | 2 - 3 | 249 | 72 | ||
| FQ214935.1 | - | 2 - 3 | 252 | 72 | ||
| FQ215084.1 | - | 2 - 3 | 179 | 72 | ||
| FQ216950.1 | - | 2 - 3 | 249 | 72 | ||
| FQ217027.1 | - | 2 - 3 | 213 | 72 | ||
| FQ217160.1 | - | 2 - 3 | 203 | 72 | ||
| FQ217559.1 | - | 2 - 3 | 202 | 72 | ||
| FQ217635.1 | - | 2 - 3 | 208 | 72 | ||
| FQ218023.1 | - | 2 - 3 | 208 | 72 | ||
| FQ218046.1 | - | 2 - 3 | 208 | 72 | ||
| FQ222358.1 | - | 2 - 3 | 236 | 72 | ||
| FQ223590.1 | - | 2 - 3 | 261 | 72 | ||
| FQ223710.1 | - | 2 - 3 | 208 | 72 | ||
| FQ223763.1 | - | 2 - 3 | 249 | 72 | ||
| FQ223794.1 | - | 2 - 3 | 254 | 72 | ||
| FQ223992.1 | - | 2 - 3 | 231 | 72 | ||
| FQ224189.1 | - | 2 - 3 | 252 | 72 | ||
| FQ224205.1 | - | 2 - 3 | 252 | 72 | ||
| FQ224455.1 | - | 2 - 3 | 251 | 72 | ||
| FQ224486.1 | - | 2 - 3 | 208 | 72 | ||
| FQ224608.1 | - | 2 - 3 | 249 | 72 | ||
| FQ224652.1 | - | 2 - 3 | 213 | 72 | ||
| FQ224692.1 | - | 2 - 3 | 209 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp5e
Genomic Map