Assay Details
| Gene Symbol: Gpr116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139110.1 | NP_620810.1 | 3 - 4 | 336 | 75 | |
| XM_006244594.2 | XP_006244656.1 | 3 - 4 | 551 | 75 | ||
| XM_006244597.2 | XP_006244659.1 | 3 - 4 | 550 | 75 | ||
| XM_008766832.1 | XP_008765054.1 | 3 - 4 | 553 | 75 | ||
| XM_008766833.1 | XP_008765055.1 | 3 - 4 | 478 | 75 | ||
| XM_008766834.1 | XP_008765056.1 | 3 - 4 | 554 | 75 | ||
| XM_008766835.1 | XP_008765057.1 | 3 - 4 | 552 | 75 | ||
| GenBank mRNA | AB019120.1 | - | 3 - 4 | 336 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P992535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58298 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P992535 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P992535 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P992535 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P992535 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700914339 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80946 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 24399 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40586 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 88470 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67809 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 88420 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5111 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80364 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700914339 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10926705 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926712 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926707 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926717 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926715 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926713 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926709 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926714 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926718 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926716 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926719 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926701 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926706 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926708 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926722 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926711 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926703 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926700 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926702 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926699 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926704 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926720 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926721 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10926698 | - | - |
| Rat Genome 230 2.0 Array | 1376767_at | - | - |
| Rat Genome 230 2.0 Array | 1388109_at | - | - |
| Rat Genome 230 2.0 Array | 1388109_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1398500_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1398500_at | - | - |
| Rat Genome 230 2.0 Array | 1376767_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376767_at | - | - |
| Affymetrix HT Rat Focus Array | 1376767_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388109_at | - | - |
| Affymetrix HT Rat Focus Array | 1388109_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0900013646.rn.1 | TC0900001656.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388109_at | - | - |
| Rat Expression Set 230 | 1376767_PM_at | - | - |
| Rat Expression Set 230 | 1388109_PM_at | - | - |
| Rat Expression Set 230 | 1376767_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10926698 | - | - |
| Rat Genome U34 Set | rc_AA900213_at | - | - |
| Rat Genome U34 Set | rc_AA997889_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21347905 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365711 | - | - |
| RatRef-12 Expression BeadChip | ILMN_61931 | - | - |

Gpr116
Genomic Map