Assay Details
| Gene Symbol: Lmbrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139189.2 | NP_631928.2 | 9 - 10 | 991 | 86 | |
| GenBank mRNA | AY093598.1 | - | 9 - 10 | 903 | 86 | |
| BC078723.1 | - | 9 - 10 | 991 | 7123105 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P451120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143798 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223596 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47752 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P451120 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P451120 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P451120 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P451120 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15367 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40741 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38385 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38046 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58785 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14382 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10163 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10922237 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922241 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922236 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922230 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922229 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922231 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922233 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922242 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922232 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922238 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922228 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922234 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922239 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922235 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922240 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922227 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10922226 | - | - |
| Rat Genome 230 2.0 Array | 1373051_at | - | - |
| Rat Genome 230 2.0 Array | 1373051_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387757_at | - | - |
| Rat Genome 230 2.0 Array | 1387757_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373051_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373051_at | - | - |
| Affymetrix HT Rat Focus Array | 1387757_at | - | - |
| Affymetrix HT Rat Focus Array | 1387757_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0900003186.rn.1 | TC0900000389.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387757_at | - | - |
| Rat Expression Set 230 | 1387757_PM_at | - | - |
| Rat Expression Set 230 | 1373051_PM_at | - | - |
| Rat Expression Set 230 | 1373051_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10922226 | - | - |
| Rat Genome U34 Set | rc_AA946120_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22041701 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358501 | - | - |
| RatRef-12 Expression BeadChip | ILMN_56536 | - | - |

Lmbrd1
Genomic Map