Assay Details
| Gene Symbol: Ctsq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139262.1 | NP_640355.1 | 2 - 3 | 146 | 90 | |
| GenBank mRNA | AF187323.1 | - | 2 - 3 | 146 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF187323.1, NM_139262.1 transcripts.
Set Membership: |
|

Ctsq
Genomic Map