Assay Details
| Gene Symbol: Rab31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145094.2 | NP_659562.2 | 6 - 7 | 582 | 81 | |
| GenBank mRNA | AF254800.1 | - | 6 - 7 | 531 | 81 | |
| BC072698.1 | - | 6 - 7 | 582 | 7105732 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P380839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478844 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532085 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601218 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131027 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89168 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P380839 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P380839 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P380839 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P380839 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P271650 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29757 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 8564 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50114 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 77363 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66565 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18295 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33435 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 26437 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21483 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58753 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10930266 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930262 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930263 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930261 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930264 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930260 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930267 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930265 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10930259 | - | - |
| Rat Genome 230 2.0 Array | 1370780_at | - | - |
| Rat Genome 230 2.0 Array | 1373894_at | - | - |
| Rat Genome 230 2.0 Array | 1370780_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1373894_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370780_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370780_at | - | - |
| Affymetrix HT Rat Focus Array | 1373894_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373894_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0900021215.rn.1 | TC0900002591.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900021214.rn.1 | TC0900002591.rn.1 | RTA TC |
| Rat Expression Set 230 | 1373894_PM_at | - | - |
| Rat Expression Set 230 | 1373894_at | - | - |
| Rat Expression Set 230 | 1370780_PM_at | - | - |
| Rat Expression Set 230 | 1370780_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930259 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20907155 | - | - |
| ABI Rat Genome Survey Microarray | 22088471 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374976 | - | - |
| RatRef-12 Expression BeadChip | ILMN_69477 | - | - |

Rab31
Genomic Map