Assay Details
| Gene Symbol: Ggta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145674.1 | NP_663707.1 | 1 - 2 | 43 | 86 | |
| GenBank mRNA | AF520589.1 | - | 1 - 2 | 43 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF520589.1, NM_145674.1 transcripts.
Set Membership: |
|

Ggta1
Genomic Map