Assay Details
| Gene Symbol: Casc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147144.1 | NP_671485.1 | 7 - 8 | 1455 | 70 | |
| GenBank mRNA | AF525467.1 | - | 7 - 8 | 1455 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P489244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172664 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P049017 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P049017 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P049017 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P489244 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 565 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27293 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10738094 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10738089 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10738096 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10738091 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10738085 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10738092 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10738088 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10738093 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10738090 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746971 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10738095 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10738087 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10738086 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10738084 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10746968 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10738083 | - | - |
| Rat Genome 230 2.0 Array | 1397571_at | - | - |
| Rat Genome 230 2.0 Array | 1397571_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1394704_at | - | - |
| Rat Genome 230 2.0 Array | 1394704_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381711_at | - | - |
| Rat Genome 230 2.0 Array | 1372089_at | - | - |
| Rat Genome 230 2.0 Array | 1372089_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381711_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1394704_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1394704_at | - | - |
| Affymetrix HT Rat Focus Array | 1397571_at | - | - |
| Affymetrix HT Rat Focus Array | 1397571_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000014137.rn.1 | TC1000001590.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000014138.rn.1 | TC1000001590.rn.1 | RTA TC |
| Rat Expression Set 230 | 1381711_PM_at | - | - |
| Rat Expression Set 230 | 1397571_at | - | - |
| Rat Expression Set 230 | 1394704_at | - | - |
| Rat Expression Set 230 | 1397571_PM_at | - | - |
| Rat Expression Set 230 | 1394704_PM_at | - | - |
| Rat Expression Set 230 | 1381711_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10738083 | - | - |
| Rat Genome U34 Set | rc_AI232353_at | - | - |
| Rat Genome U34 Set | rc_AI234847_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22165548 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352423 | - | - |
| RatRef-12 Expression BeadChip | ILMN_59077 | - | - |

Casc3
Genomic Map