Assay Details
| Gene Symbol: Ly49i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152848.1 | NP_690061.1 | 1 - 2 | 119 | 143 | |
| GenBank mRNA | AY115572.1 | - | 1 - 2 | 119 | 143 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY115572.1, NM_152848.1 transcripts.
Set Membership: |
|

Ly49i2
Genomic Map