Assay Details
| Gene Symbol: Ccnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171992.4 | NP_741989.3 | 2 - 3 | 541 | 87 | |
| XM_008760168.1 | XP_008758390.1 | 2 - 3 | 716 | 87 | ||
| GenBank mRNA | CA510842.1 | - | 2 - 3 | 542 | 87 | |
| D14014.1 | - | 2 - 3 | 541 | 87 | ||
| FQ231654.1 | - | 2 - 3 | 554 | 87 | ||
| X75207.1 | - | 2 - 3 | 568 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccnd1
Genomic Map