Assay Details
| Gene Symbol: Fzd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172035.1 | NP_742032.1 | 1 | 748 | 121 | |
| GenBank mRNA | L02530.1 | - | 1 | 748 | 121 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fzd2
Genomic Map