Assay Details
| Gene Symbol: Pcyt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173151.1 | NP_775174.1 | 5 - 6 | 571 | 72 | |
| XM_006257016.2 | XP_006257078.1 | 5 - 6 | 532 | 72 | ||
| XM_006257017.2 | XP_006257079.1 | 5 - 6 | 939 | 72 | ||
| XM_008773254.1 | XP_008771476.1 | 5 - 6 | 652 | 72 | ||
| GenBank mRNA | AF190256.1 | - | 5 - 6 | 571 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P095201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114662 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P095201 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P095201 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P095201 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P418995 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16458 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30797 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66190 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 56152 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10933985 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933989 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933993 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933992 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933994 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933990 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933987 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933991 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933988 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933986 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10933984 | - | - |
| Rat Genome 230 2.0 Array | 1370700_at | - | - |
| Rat Genome 230 2.0 Array | 1370700_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370700_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370700_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0X00003298.rn.1 | TC0X00000526.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370700_PM_at | - | - |
| Rat Expression Set 230 | 1370700_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933984 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20908741 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357159 | - | - |
| RatRef-12 Expression BeadChip | ILMN_60774 | - | - |

Pcyt1b
Genomic Map