Assay Details
| Gene Symbol: Eno4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134505.1 | NP_001127977.1 | 9 - 10 | 1238 | 62 | |
| XM_006231669.2 | XP_006231731.1 | 9 - 10 | 7854 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eno4
Genomic Map