Assay Details
| Gene Symbol: Tacr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012667.2 | NP_036799.1 | 941 | 69 | ||
| GenBank mRNA | J05097.1 | - | 941 | 69 | ||
| M31477.1 | - | 1 - 2 | 398 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tacr1
Genomic Map