Assay Details
| Gene Symbol: Tsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021762.1 | NP_068530.1 | 1 - 2 | 177 | 100 | |
| XM_008769496.1 | XP_008767718.1 | 1 - 2 | 196 | 100 | ||
| GenBank mRNA | AF262356.1 | - | 1 - 2 | 177 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tsn
Genomic Map