Assay Details
| Gene Symbol: Hhex | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024385.1 | NP_077361.1 | 2 - 3 | 552 | 64 | |
| GenBank mRNA | BC088135.1 | - | 2 - 3 | 557 | 7366155 | 64 |
| D86383.1 | - | 2 - 3 | 552 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hhex
Genomic Map