Assay Details
| Gene Symbol: Rabep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030585.1 | NP_085074.1 | 11 - 12 | 1511 | 70 | |
| GenBank mRNA | BC089795.1 | - | 11 - 12 | 1497 | 7375879 | 70 |
| U34932.1 | - | 11 - 12 | 1511 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rabep2
Genomic Map