Assay Details
| Gene Symbol: Sv2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031593.1 | NP_113781.1 | 6 - 7 | 1360 | 61 | |
| GenBank mRNA | AF060174.1 | - | 6 - 7 | 1360 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sv2c
Genomic Map