Assay Details
| Gene Symbol: Pla2g2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031598.3 | NP_113786.3 | 2 - 3 | 104 | 79 | |
| XM_006239146.2 | XP_006239208.1 | 3 - 4 | 270 | 79 | ||
| GenBank mRNA | AF365363.1 | - | 1 - 2 | 48 | 79 | |
| AY651027.1 | - | 1 - 2 | 48 | 79 | ||
| D00523.1 | - | 2 - 3 | 88 | 79 | ||
| FQ227448.1 | - | 2 - 3 | 104 | 79 | ||
| FQ234749.1 | - | 3 - 4 | 202 | 79 | ||
| M25148.1 | - | 2 - 3 | 104 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pla2g2a
Genomic Map