Assay Details
| Gene Symbol: Zfp148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031615.1 | NP_113803.1 | 7 - 8 | 1047 | 65 | |
| XM_006248435.2 | XP_006248497.1 | 4 - 5 | 695 | 65 | ||
| GenBank mRNA | U30381.1 | - | 7 - 8 | 1047 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
protein complex assembly
gamete generation
negative regulation of gene expression
substantia nigra development
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
transcription regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
metal ion binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
protein complex assembly
gamete generation
negative regulation of gene expression
substantia nigra development
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
transcription regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P146969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8033 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P146969 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P146969 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P146969 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P146969 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702022982 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19887 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 25054 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46381 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18929 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17710 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57120 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17923 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50502 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 702022982 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10754637 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754639 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754631 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754638 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754634 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754636 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754630 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754633 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754632 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754640 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754635 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10754629 | - | - |
| Rat Genome 230 2.0 Array | 1381998_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381998_at | - | - |
| Rat Genome 230 2.0 Array | 1368067_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368067_at | - | - |
| Affymetrix HT Rat Focus Array | 1368067_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368067_at | - | - |
| Affymetrix HT Rat Focus Array | 1381998_at | - | - |
| Affymetrix HT Rat Focus Array | 1381998_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1100010756.rn.1 | TC1100001736.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100010757.rn.1 | TC1100001736.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100010759.rn.1 | TC1100001736.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368067_PM_at | - | - |
| Rat Expression Set 230 | 1368067_at | - | - |
| Rat Expression Set 230 | 1381998_PM_at | - | - |
| Rat Expression Set 230 | 1381998_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754629 | - | - |
| Rat Genome U34 Set | U30381_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21225828 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363968 | - | - |
| RatRef-12 Expression BeadChip | ILMN_53875 | - | - |

Zfp148
Genomic Map