Assay Details
| Gene Symbol: Klf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031642.4 | NP_113830.2 | 2 - 3 | 920 | 68 | |
| XM_006254182.1 | XP_006254244.1 | 9 - 10 | 2186 | 68 | ||
| GenBank mRNA | AF001417.1 | - | 2 - 3 | 700 | 68 | |
| AY318957.1 | - | 9 - 10 | 2185 | 68 | ||
| BC097306.1 | - | 2 - 3 | 845 | 7444817 | 68 | |
| FQ225029.1 | - | 1 - 2 | 476 | 68 | ||
| FQ230654.1 | - | 1 - 2 | 570 | 68 | ||
| FQ231202.1 | - | 1 - 2 | 260 | 68 | ||
| FQ231451.1 | - | 1 - 2 | 260 | 68 | ||
| FQ232231.1 | - | 1 - 2 | 259 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klf6
Genomic Map