Assay Details
| Gene Symbol: Slco1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270586.1 | NP_001257515.1 | 4 - 5 | 474 | 68 | |
| NM_001270587.1 | NP_001257516.1 | 4 - 5 | 474 | 68 | ||
| NM_031650.3 | NP_113838.1 | 4 - 5 | 474 | 68 | ||
| GenBank mRNA | AF147740.1 | - | 3 - 4 | 360 | 68 | |
| AF208545.2 | - | 4 - 5 | 474 | 68 | ||
| AF217450.2 | - | 4 - 5 | 474 | 68 | ||
| AJ271682.1 | - | 4 - 5 | 475 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slco1b2
Genomic Map