Assay Details
| Gene Symbol: Slc16a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031744.2 | NP_113932.2 | 4 - 5 | 565 | 71 | |
| XM_006242039.2 | XP_006242101.1 | 3 - 4 | 834 | 71 | ||
| GenBank mRNA | AF059258.1 | - | 4 - 5 | 565 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc16a8
Genomic Map