Assay Details
| Gene Symbol: Cnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031747.1 | NP_113935.1 | 2 - 3 | 249 | 63 | |
| GenBank mRNA | BC061809.1 | - | 2 - 3 | 218 | 5598777 | 63 |
| D14437.1 | - | 2 - 3 | 249 | 63 | ||
| X71071.1 | - | 2 - 3 | 214 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnn1
Genomic Map