Assay Details
| Gene Symbol: NEWGENE_621351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762736.1 | XP_008760958.1 | 33 - 34 | 2396 | 103 | |
| Gene Symbol: Col1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053356.1 | NP_445808.1 | 2046 | 103 | ||
| GenBank mRNA | AF121217.1 | - | 2046 | 103 | ||
| BC108298.1 | - | 2139 | 103 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

NEWGENE_621351
Genomic Map