Assay Details
| Gene Symbol: LOC100911034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749609.2 | XP_003749657.1 | 11 - 12 | 1249 | 79 | |
| XM_006235451.2 | XP_006235513.1 | 11 - 12 | 1243 | 79 | ||
| Gene Symbol: Nfs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053462.2 | NP_445914.2 | 1244 | 79 | ||
| XM_006235365.2 | XP_006235427.1 | 1243 | 79 | |||
| GenBank mRNA | AF336041.1 | - | 1205 | 79 | ||
| BC104699.1 | - | 1244 | 7189705 | 79 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911034
Genomic Map