Assay Details
| Gene Symbol: LOC100911034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749609.2 | XP_003749657.1 | 5 - 6 | 589 | 67 | |
| XM_006235451.2 | XP_006235513.1 | 5 - 6 | 583 | 67 | ||
| Gene Symbol: Nfs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053462.2 | NP_445914.2 | 584 | 67 | ||
| XM_006235365.2 | XP_006235427.1 | 583 | 67 | |||
| GenBank mRNA | AF336041.1 | - | 545 | 67 | ||
| BC089205.1 | - | 550 | 67 | |||
| BC104699.1 | - | 584 | 7189705 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC100911034
Genomic Map