Assay Details
| Gene Symbol: Nme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053507.1 | NP_445959.1 | 3 - 4 | 276 | 90 | |
| XM_008767598.1 | XP_008765820.1 | 3 - 4 | 388 | 90 | ||
| GenBank mRNA | AY017337.1 | - | 3 - 4 | 276 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nme3
Genomic Map