Assay Details
| Gene Symbol: Prdx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053512.2 | NP_445964.1 | 1 - 2 | 272 | 72 | |
| GenBank mRNA | AF106945.1 | - | 1 - 2 | 273 | 72 | |
| BC059122.1 | - | 1 - 2 | 356 | 6918063 | 72 | |
| FQ219258.1 | - | 1 - 2 | 320 | 72 | ||
| FQ220216.1 | - | 1 - 2 | 326 | 72 | ||
| FQ220257.1 | - | 1 - 2 | 320 | 72 | ||
| FQ220425.1 | - | 1 - 2 | 321 | 72 | ||
| FQ220916.1 | - | 1 - 2 | 321 | 72 | ||
| FQ223235.1 | - | 1 - 2 | 316 | 72 | ||
| FQ228535.1 | - | 1 - 2 | 321 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Prdx4
Genomic Map