Assay Details
| Gene Symbol: Rax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053678.1 | NP_446130.1 | 1 - 2 | 283 | 89 | |
| GenBank mRNA | AF135839.1 | - | 1 - 2 | 283 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rax
Genomic Map