Assay Details
| Gene Symbol: Polr2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053948.4 | NP_446400.1 | 1 - 2 | 83 | 72 | |
| XM_006230973.1 | XP_006231035.1 | 3 - 4 | 368 | 72 | ||
| GenBank mRNA | BC060550.1 | - | 1 - 2 | 39 | 6890594 | 72 |
| BC060595.1 | - | 3 - 4 | 377 | 6888231 | 72 | |
| FQ223906.1 | - | 3 - 4 | 373 | 72 | ||
| FQ224086.1 | - | 1 - 2 | 116 | 72 | ||
| FQ232354.1 | - | 1 - 2 | 87 | 72 | ||
| Z71925.1 | - | 1 - 2 | 84 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC060550.1, BC060595.1, NM_053948.4, XM_006230973.1, Z71925.1 transcripts.
Set Membership: |
|

Polr2g
Genomic Map