Assay Details
| Gene Symbol: Guca2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022284.2 | NP_071620.1 | 1 - 2 | 111 | 61 | |
| GenBank mRNA | U41322.1 | - | 1 - 2 | 111 | 61 | |
| U73898.1 | - | 1 - 2 | 127 | 61 | ||
| U75186.1 | - | 1 - 2 | 238 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Guca2b
Genomic Map