Assay Details
| Gene Symbol: Des | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022531.1 | NP_071976.1 | 6 - 7 | 1320 | 79 | |
| GenBank mRNA | BC061872.1 | - | 6 - 7 | 1313 | 5598951 | 79 |
| FQ216012.1 | - | 2 - 3 | 286 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Des
Genomic Map