Assay Details
| Gene Symbol: Araf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033663.2 | NP_001028835.1 | 5 - 6 | 571 | 102 | |
| NM_022532.2 | NP_071977.1 | 5 - 6 | 571 | 102 | ||
| XM_008773036.1 | XP_008771258.1 | 5 - 6 | 1313 | 102 | ||
| GenBank mRNA | AB158253.1 | - | 4 - 5 | 467 | 102 | |
| BC060568.1 | - | 611 | 102 | |||
| FQ213741.1 | - | 1 - 2 | 143 | 102 | ||
| X06942.1 | - | 4 - 5 | 538 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPKK activity
protein phosphorylation
regulation of TOR signaling
regulation of proteasomal ubiquitin-dependent protein catabolic process
positive regulation of peptidyl-serine phosphorylation
intracellular signal transduction
negative regulation of apoptotic process
protein serine/threonine kinase activity
MAP kinase kinase kinase activity
receptor signaling protein activity
ATP binding
metal ion binding
MAPK cascade
activation of MAPKK activity
protein phosphorylation
regulation of TOR signaling
regulation of proteasomal ubiquitin-dependent protein catabolic process
positive regulation of peptidyl-serine phosphorylation
intracellular signal transduction
negative regulation of apoptotic process
protein serine/threonine kinase activity
MAP kinase kinase kinase activity
receptor signaling protein activity
ATP binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P150691 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P150699 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P109880 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P150696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152377 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P109880 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P150691 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P150696 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P150699 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P150691 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P109880 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P150699 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P150696 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P150696 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P150699 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P150691 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P109880 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P322960 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P211440 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700506813 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 61217 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36981 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55217 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 26774 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 60757 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 61889 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 69563 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10413 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38446 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14052 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59417 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5117 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700506813 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10936495 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936492 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936494 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936490 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936503 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936501 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936497 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936507 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936500 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936508 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936491 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936499 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936493 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936502 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936504 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936505 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936496 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936506 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936489 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936509 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10936488 | - | - |
| Rat Genome 230 2.0 Array | 1390196_at | - | - |
| Rat Genome 230 2.0 Array | 1368390_at | - | - |
| Rat Genome 230 2.0 Array | 1390196_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368390_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388305_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388305_at | - | - |
| Affymetrix HT Rat Focus Array | 1388305_at | - | - |
| Affymetrix HT Rat Focus Array | 1368390_at | - | - |
| Affymetrix HT Rat Focus Array | 1368390_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388305_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0X00008585.rn.1 | TC0X00001372.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00008587.rn.1 | TC0X00001372.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00008584.rn.1 | TC0X00001372.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00008588.rn.1 | TC0X00001372.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00008586.rn.1 | TC0X00001372.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00008589.rn.1 | TC0X00001372.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388305_PM_at | - | - |
| Rat Expression Set 230 | 1368390_PM_at | - | - |
| Rat Expression Set 230 | 1388305_at | - | - |
| Rat Expression Set 230 | 1368390_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10936488 | - | - |
| Rat Genome U34 Set | X06942_at | - | - |
| Rat Genome U34 Set | rc_AI172412_at | - | - |
| Rat Genome U34 Set | rc_AI227665_s_at | - | - |
| Rat Genome U34 Set | X06942_at | - | - |
| Rat Genome U34 Set | rc_AI227665_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352852 | - | - |
| RatRef-12 Expression BeadChip | ILMN_66915 | - | - |

Araf
Genomic Map