Assay Details
| Gene Symbol: Pdpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031081.1 | NP_112343.1 | 6 - 7 | 778 | 66 | |
| XM_006246036.2 | XP_006246098.1 | 6 - 7 | 2630 | 66 | ||
| XM_006246038.2 | XP_006246100.1 | 6 - 7 | 840 | 66 | ||
| GenBank mRNA | Y15748.1 | - | 6 - 7 | 778 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdpk1
Genomic Map