Assay Details
| Gene Symbol: Csn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031562.1 | NP_113750.1 | 3 - 4 | 137 | 134 | |
| GenBank mRNA | K02598.1 | - | 3 - 4 | 137 | 134 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the K02598.1, NM_031562.1 transcripts.
Set Membership: |
|

Csn3
Genomic Map