Assay Details
| Gene Symbol: Cryba4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031689.1 | NP_113877.1 | 2 - 3 | 80 | 60 | |
| GenBank mRNA | AF013247.1 | - | 2 - 3 | 80 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cryba4
Genomic Map