Assay Details
| Gene Symbol: Clic4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031818.1 | NP_114006.1 | 3 - 4 | 311 | 99 | |
| GenBank mRNA | AF104119.1 | - | 3 - 4 | 311 | 99 | |
| EF397567.1 | - | 3 - 4 | 311 | 99 | ||
| FQ222737.1 | - | 421 | 99 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Clic4
Genomic Map