Assay Details
| Gene Symbol: Clcnka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053327.1 | NP_445779.1 | 4 - 5 | 396 | 70 | |
| XM_008764291.1 | XP_008762513.1 | 4 - 5 | 512 | 70 | ||
| XM_008764292.1 | XP_008762514.1 | 4 - 5 | 482 | 70 | ||
| GenBank mRNA | BC081761.1 | - | 4 - 5 | 462 | 7108273 | 70 |
| D13927.1 | - | 4 - 5 | 396 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clcnka
Genomic Map