Assay Details
| Gene Symbol: Gipc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053341.1 | NP_445793.1 | 1 - 2 | 344 | 63 | |
| XM_006255311.2 | XP_006255373.1 | 1 - 2 | 2609 | 63 | ||
| XM_006255312.2 | XP_006255374.1 | 2 - 3 | 1603 | 63 | ||
| XM_008772420.1 | XP_008770642.1 | 2 - 3 | 1533 | 63 | ||
| GenBank mRNA | AF032120.1 | - | 1 - 2 | 344 | 63 | |
| AF089817.1 | - | 2 - 3 | 362 | 63 | ||
| BC070505.1 | - | 2 - 3 | 387 | 7097697 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gipc1
Genomic Map