Assay Details
| Gene Symbol: Lrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053541.1 | NP_445993.1 | 4 - 5 | 845 | 74 | |
| XM_008759197.1 | XP_008757419.1 | 3 - 4 | 2005 | 74 | ||
| XM_008759198.1 | XP_008757420.1 | 3 - 4 | 2005 | 74 | ||
| XM_008759199.1 | XP_008757421.1 | 3 - 4 | 2005 | 74 | ||
| XM_008759200.1 | XP_008757422.1 | 4 - 5 | 924 | 74 | ||
| XM_008759201.1 | XP_008757423.1 | 3 - 4 | 2004 | 74 | ||
| XM_008759202.1 | XP_008757424.1 | 4 - 5 | 924 | 74 | ||
| GenBank mRNA | AB009463.1 | - | 4 - 5 | 845 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrp3
Genomic Map