Assay Details
| Gene Symbol: Dut | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053592.2 | NP_446044.1 | 1 - 2 | 162 | 110 | |
| XM_008762188.1 | XP_008760410.1 | 1 - 2 | 200 | 110 | ||
| GenBank mRNA | U64030.2 | - | 1 - 2 | 162 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dut
Genomic Map