Assay Details
| Gene Symbol: Mmp23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053606.2 | NP_446058.1 | 5 - 6 | 778 | 82 | |
| GenBank mRNA | AB010960.1 | - | 5 - 6 | 939 | 82 | |
| BC086586.1 | - | 5 - 6 | 778 | 7309721 | 82 | |
| FQ220321.1 | - | 5 - 6 | 784 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmp23
Genomic Map